Anti-OTUB2 Mouse Monoclonal Antibody [clone: 6C7]
GNTXGTX83952
: Genetex
- Antibody type:Primary
- Antigen name:OTU deubiquitinase, ubiquitin aldehyde binding 2
- Antigen symbol:OTUB2
- Clonality:Monoclonal
- Clone:6C7
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Gel shift:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Gene ID:78990
- Antigen synonyms:C14orf137|OTU2|OTB2
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Storage temperature:For short-term storage, store at 4 °C or store at –20 °C. For long-term storage, store at -70°C (aliquotted). Avoid repeated freeze-thaw cycles.
- Immunogen:Full length human recombinant protein of human OTUB2 (NP_075601) produced in HEK293T cell.
- Purification:Purified from mouse ascites fluids by Protein A/G affinity chromatography
- Size:100 μl
- Pk:100 µl
Type: Primary
Antigen: OTUB2
Clonality: Monoclonal
Clone: Unconjugated
Conjugation: 6C7
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human