Anti-PYCR2 Mouse Monoclonal Antibody [clone: 4A10]
Catalog # GNTXGTX83752
Supplier: Genetex
Specifications
- Antibody type:Primary
- Antigen name:Pyrroline-5-carboxylate reductase 2
- Antigen symbol:PYCR2
- Clonality:Monoclonal
- Clone:4A10
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Gel shift:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG2a
- Reactivity:Human
- Western blot:Yes
- Gene ID:29920
- Antigen synonyms:HLD10|P5CR2
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Storage temperature:For short-term storage, store at 4 °C or store at –20 °C. For long-term storage, store at -70°C (aliquotted). Avoid repeated freeze-thaw cycles.
- Immunogen:Full length human recombinant protein of human PYCR2 (NP_037460) produced in HEK293T cell.
- Purification:Purified from mouse ascites fluids by Protein A/G affinity chromatography
- Size:100 μl
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: PYCR2
Clonality: Monoclonal
Clone: Unconjugated
Conjugation: 4A10
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human