Anti-RAB17 Mouse Monoclonal Antibody [clone: 5E2]
Catalog # GNTXGTX83741
Supplier: Genetex
Specifications
- Antibody type:Primary
- Antigen name:Ras-related protein Rab-17
- Antigen symbol:RAB17
- Clonality:Monoclonal
- Clone:5E2
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Gene ID:64284
- Antigen synonyms:BOFS|AP2TF|AP-2|AP-2alpha|TFAP2
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Storage temperature:For short-term storage, store at 4 °C or store at –20 °C. For long-term storage, store at -70°C (aliquotted). Avoid repeated freeze-thaw cycles.
- Immunogen:Full length human recombinant protein of human RAB17(NP_071894) produced in HEK293T cell.
- Purification:Purified from mouse ascites fluids by Protein A/G affinity chromatography
- Size:100 μl
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: RAB17
Clonality: Monoclonal
Clone: Unconjugated
Conjugation: 5E2
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human