Anti-IL6 Mouse Monoclonal Antibody [clone: 6D9A1,6C12B1]
Référence GNTXGTX83394
Fournisseur: Genetex
Spécifications
- Antibody type:Primary
- Antigen name:Interleukin 6
- Antigen symbol:IL6
- Clonality:Monoclonal
- Clone:6D9A1,6C12B1
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Gene ID:3569
- Antigen synonyms:HSF|HGF|IFNB2|interferon beta 2|BSF2|IL-6
- Storage buffer:Ascitic fluid containing 0,03% sodium azide.
- Storage temperature:Store at 4℃, for long term storage, store at -20℃.
- Immunogen:The immunogen corresponding to human IL-6 expressed in E. Coli.
- Size:100 μl
- Pk:100 µl
Spécifications
À propos de cet article
Type: Primary
Antigen: IL6
Clonality: Monoclonal
Clone: Unconjugated
Conjugation: 6D9A1,6C12B1
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human