Anti-MSH2 Mouse Monoclonal Antibody [clone: 1B3/3A2B8C]
Katalognr. GNTXGTX83390
Supplier: Genetex
Specifications
- Antibody type:Primary
- Antigen name:MutS homolog 2
- Antigen symbol:MSH2
- Clonality:Monoclonal
- Clone:1B3/3A2B8C
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Monkey,Human
- Western blot:Yes
- Gene ID:4436
- Antigen synonyms:DNA mismatch repair protein Msh2|COCA1|FCC1|HNPCC|hMSH2|HNPCC1|LCFS2
- Storage buffer:Ascitic fluid containing 0,03% sodium azide.
- Storage temperature:Store at 4℃, for long term storage, store at -20℃.
- Immunogen:The immunogen corresponding to human MSH2 expressed in E. Coli.
- Size:100 μl
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: MSH2
Clonality: Monoclonal
Clone: Unconjugated
Conjugation: 1B3/3A2B8C
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Monkey