Anti-C-Kit (v-kit Hardy-Zuckerman 4 feline sarcoma viral oncogene homolog) Mouse Monoclonal Antibody [clone: 8D7]
GNTXGTX83074
: Genetex
- Antibody type:Primary
- Antigen name:Proto-oncogene Receptor Tyrosine Kinase
- Antigen symbol:KIT
- Clonality:Monoclonal
- Clone:8D7
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Gene ID:3815
- Antigen synonyms:SCFR|CD117|PBT|C-Kit
- Storage buffer:Ascitic fluid containing 0,03% sodium azide.
- Storage temperature:Store at 4℃, for long term storage, store at -20℃.
- Immunogen:The immunogen corresponding to c-Kit.
- Size:100 μl
- Pk:100 µl
Type: Primary
Antigen: C-Kit (v-kit Hardy-Zuckerman 4 feline sarcoma viral oncogene homolog)
Clonality: Monoclonal
Clone: Unconjugated
Conjugation: 8D7
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human