Anti-ABL2 Mouse Monoclonal Antibody [clone: 1H1]
GNTXGTX82788
: Genetex
- Antibody type:Primary
- Antigen name:ABL proto-oncogene 2, Non-receptor Tyrosine Kinase
- Antigen symbol:ABL2
- Clonality:Monoclonal
- Clone:1H1
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Human,Mouse
- Western blot:Yes
- Gene ID:27
- Antigen synonyms:ARG, ABLL, FLJ22224, FLJ31718, FLJ41441, ABL2
- Storage buffer:Ascitic fluid containing 0,03% sodium azide.
- Storage temperature:Store at 4℃, for long term storage, store at -20℃.
- Immunogen:The immunogen corresponding to ABL2 expressed in E. Coli.
- Size:100 μl
- Pk:100 µl
Type: Primary
Antigen: ABL2
Clonality: Monoclonal
Clone: Unconjugated
Conjugation: 1H1
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse