Anti-c-Myc (v-myc myelocytomatosis viral oncogene homolog (avian)) Mouse Monoclonal Antibody (FITC (Fluorescein isothiocyanate)) [clone: 9E10]
Catalog # GNTXGTX80250
Supplier: Genetex
Specifications
- Antibody type:Primary
- Antigen symbol:c-Myc (v-myc myelocytomatosis viral oncogene homolog (avian))
- Clonality:Monoclonal
- Clone:9E10
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Reactivity:Human
- Western blot:Yes
- Size:100 μg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: c-Myc (v-myc myelocytomatosis viral oncogene homolog (avian))
Clonality: Monoclonal
Clone: FITC (Fluorescein isothiocyanate)
Conjugation: 9E10
Epitope:
Host: Mouse
Isotype:
Reactivity: Human