Order Entry
Switzerland
ContactUsLinkComponent
Anti-CD66 molecule Mouse Monoclonal Antibody (CF568) [clone: C66/261]
Anti-CD66 molecule Mouse Monoclonal Antibody (CF568) [clone: C66/261]
Supplier:  Biotium
Certificates
Anti-CD66 molecule Mouse Monoclonal Antibody (CF568) [clone: C66/261]
Supplier:  Biotium

Specifications

  • Catalog No:
  • BNC680261-100
  • BNC680261-500
  • Antigen symbol:
  • CD66 molecule
  • CD66 molecule
  • Antigen name:
  • CD66
  • CD66
  • Antigen synonyms:
  • CEACAM5|CD66|Biliary Glycoprotein (BGP-1)|Carcinoembryonic Antigen-related Cell Adhesion Molecule 5
  • CEACAM5|CD66|Biliary Glycoprotein (BGP-1)|Carcinoembryonic Antigen-related Cell Adhesion Molecule 5
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 568
  • CF® 568
  • Clone:
  • Clone C66/261
  • Clone C66/261
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 709196
  • 709196
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

Specifications

Frequently Bought Together