Anti-CD264 Mouse Monoclonal Antibody (Phycoerythrin (PE)) [clone: TRAIL-R4-01]
Catalog # GNTXGTX79876
Supplier: Genetex
Specifications
- Antibody type:Primary
- Antigen name:Tumor Necrosis Factor Receptor Superfamily, Member 10D, Decoy With Truncated Death Domain
- Antigen symbol:CD264
- Clonality:Monoclonal
- Clone:TRAIL-R4-01
- Conjugation:PE (Phycoerythrin)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Gene ID:8793
- Antigen synonyms:TRAILR4|TRAIL-R4|CD264|TRUNDD|DCR2
- Storage buffer:The reagent is provided in phosphate buffered saline (PBS) containing 15 mM sodium azide and 0,2% (w/v) high-grade protease free Bovine Serum Albumin (BSA) as a stabilizing agent.
- Storage temperature:Store in the dark at 2-8 °C. Do not freeze. Do not use after expiration date stamped on vial label. Short-term exposure to room temperature should not affect the quality of the reagent.
- Immunogen:TRAIL-R4 (aa 1-210) - hIgGhc fusion protein
- Size:100 μg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: CD264
Clonality: Monoclonal
Clone: Phycoerythrin (PE)
Conjugation: TRAIL-R4-01
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human