Anti-REN Mouse Monoclonal Antibody (FITC (Fluorescein isothiocyanate)) [clone: 4B5-E3]
Catalog # GNTXGTX79680
Supplier: Genetex
Specifications
- Antibody type:Primary
- Antigen name:Renin
- Antigen symbol:REN
- Clonality:Monoclonal
- Clone:4B5-E3
- Conjugation:FITC (Fluorescein Isothiocyanate)
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2b
- Reactivity:Human
- Western blot:Yes
- Gene ID:5972
- Antigen synonyms:renin precursor|HNFJ2|angiotensinogenase|renal|angiotensin-forming enzyme
- Storage buffer:0,05M Sodium Phosphate
- Storage temperature:Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid multiple freeze-thaw cycles.
- Concentration:1 mg/ml
- Size:50 μg
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: REN
Clonality: Monoclonal
Clone: FITC (Fluorescein isothiocyanate)
Conjugation: 4B5-E3
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human