Anti-CNG Channel (cyclic nucleotide gated channel alpha 1) Mouse Monoclonal Antibody [clone: CNC 9C1]
Catalog # GNTXGTX79456
Supplier: Genetex
Specifications
- Antibody type:Primary
- Antigen name:Cyclic Nucleotide Gated Channel Alpha 1
- Antigen symbol:CNG Channel (cyclic nucleotide gated channel alpha 1)
- Clonality:Monoclonal
- Clone:CNC 9C1
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Gene ID:1259
- Antigen synonyms:CNCG1|RCNCalpha|RP49|RCNC1|RCNCa|CNG-1|CNCG|CNG1
- Storage buffer:Ascites containing 0,05 % sodium azide
- Storage temperature:-20° C, Avoid Freeze/Thaw Cycles
- Concentration:1 mg/ml
- Immunogen:MBP-fusion protein of the C-terminal region of the a2 subunit (amino acids K581-D670)
- Size:100 μl
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: CNG Channel (cyclic nucleotide gated channel alpha 1)
Clonality: Monoclonal
Clone: Unconjugated
Conjugation: CNC 9C1
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat