Anti-CD1A Mouse Monoclonal Antibody (FITC (Fluorescein isothiocyanate)) [clone: NA1/34-HLK]
GNTXGTX76535
: Genetex
- Antibody type:Primary
- Antigen name:CD1a molecule
- Antigen symbol:CD1A
- Clonality:Monoclonal
- Clone:NA1/34-HLK
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Cynomolgus monkey,Human,Dog
- Gene ID:909
- Antigen synonyms:FCB6|cortical thymocyte antigen CD1A|T-cell surface antigen T6/Leu-6|cluster of differentiation 1 A|T6|CD1|R4|a polypeptide|T-cell surface glycoprotein CD1a|CD1A antigen|HTA1
- Storage buffer:Phosphate buffered saline containing 0,09% Sodium Azide 1% Bovine Serum Albumin
- Storage temperature:Store at +4 °C or at –20 °C if preferred. Store undiluted. Storage in frost-free freezers is not recommended. Photosensitive. Should this product contain a precipitate we recommend microcentrifugation before use.
- Purification:Purified IgG prepared by affinity chromatography on Protein G.
- Size:100 μg
- Pk:100 µG
Type: Primary
Antigen: CD1A
Clonality: Monoclonal
Clone: FITC (Fluorescein isothiocyanate)
Conjugation: NA1/34-HLK
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Dog, ClMonkey