Anti-CD41 (Integirn alpha 2b (platelet glycoprotein IIb of IIb/IIIa Complex Antigen CD41)) Mouse Monoclonal Antibody [clone: PM6/248]
Catalog # GNTXGTX76177
Supplier: Genetex
Specifications
- Antibody type:Primary
- Antigen name:Integrin Alpha 2 Beta
- Antigen symbol:ITGA2B
- Clonality:Monoclonal
- Clone:PM6/248
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Baboon,Mustelid,Human,Mink
- Gene ID:3674
- Antigen synonyms:PPP1R93|GP2B|GTA|CD41B|HPA3|GPIIb|BDPLT2|CD41|BDPLT16|GT
- Storage buffer:Phosphate buffered saline containing 0,09% Sodium Azide
- Storage temperature:Store at +4 °C or at –20 °C if preferred. This product should be stored undiluted. Storage in frost free freezers is not recommended. Should this product contain a precipitate we recommend microcentrifugation before use.
- Purification:Purified IgG prepared by affinity chromatography on Protein G from tissue culture supernatant
- Size:200 μg
- Pk:200 µG
Specifications
About this item
Type: Primary
Antigen: CD41 (Integirn alpha 2b (platelet glycoprotein IIb of IIb/IIIa Complex Antigen CD41))
Clonality: Monoclonal
Clone: Unconjugated
Conjugation: PM6/248
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Bb, Mink, Mustelid