Anti-SLAM Mouse Monoclonal Antibody (FITC (Fluorescein isothiocyanate)) [clone: A12]
Catalog # GNTXGTX76015
Supplier: Genetex
Specifications
- Antibody type:Primary
- Antigen name:Signaling Lymphocytic Activation Molecule
- Antigen symbol:SLAM
- Clonality:Monoclonal
- Clone:A12
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Cynomolgus monkey,Human
- Gene ID:6504
- Antigen synonyms:CD150|CDw150|IPO-3|signaling lymphocytic activation molecule|SLAMF1|SLAM
- Storage buffer:Phosphate buffered saline containing 0,09% Sodium Azide 1% Bovine Serum Albumin
- Storage temperature:Store at +4 °C or at –20 °C if preferred. Store undiluted. Storage in frost-free freezers is not recommended. Photosensitive. Should this product contain a precipitate we recommend microcentrifugation before use.
- Purification:Purified IgG prepared by affinity chromatography on Protein G from tissue culture supernatant
- Size:100 μg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: SLAM
Clonality: Monoclonal
Clone: FITC (Fluorescein isothiocyanate)
Conjugation: A12
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, ClMonkey