Anti-IgL Mouse Monoclonal Antibody (FITC (Fluorescein isothiocyanate)) [clone: Mc24-IC6]
Catalog # GNTXGTX75175
Supplier: Genetex
Specifications
- Antibody type:Primary
- Antigen name:Immunoglobulin L
- Antigen symbol:IgL
- Clonality:Monoclonal
- Clone:Mc24-IC6
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Epitope:Light Chain
- Gene ID:28815
- Antigen synonyms:lambda light chain|Ig lambda|Ig lambda light chain|Immunoglobulin lambda light chain|Igl
- Storage buffer:Phosphate buffered saline containing 0,09% Sodium Azide 1% Bovine Serum Albumin
- Storage temperature:Store at +4 °C for one month or at –20 °C for longer. This product should be stored undiluted. This product is photosensitive and should be protected from light. Should this product contain a precipitate we recommend microcentrifugation before use.
- Immunogen:Purified human IgG.
- Purification:Purified IgG prepared by affinity chromatography on Protein A
- Size:100 μg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: IgL
Clonality: Monoclonal
Clone: FITC (Fluorescein isothiocyanate)
Conjugation: Mc24-IC6
Epitope: Light Chain
Host: Mouse
Isotype: IgG1
Reactivity: Human