Mouse IgG2a Isotype Control [clone: F10-89-4] (Biotin)
Catalog # GNTXGTX74755
Supplier: Genetex
Specifications
- Antibody type:Control
- Clonality:Monoclonal
- Clone:F10-89-4
- Conjugation:Biotin
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG2a
- Storage buffer:Phosphate buffered saline containing 0,09% Sodium Azide 1% Bovine Serum Albumin
- Storage temperature:Store at +4 °C or at –20 °C if preferred. This product should be stored undiluted. Storage in frost free freezers is not recommended. Should this product contain a precipitate we recommend microcentrifugation before use.
- Purification:Purified IgG prepared by ion exchange chromatography
- Size:100 μg
- Pk:100 µG
Specifications
About this item
Type: Control
Antigen: IgG2a
Clonality: Monoclonal
Clone: Biotin
Conjugation: F10-89-4
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: