Anti-PRF1 Rabbit Polyclonal Antibody
GNTXGTX59812
: Genetex
- Antibody type:Primary
- Antigen name:Perforin
- Antigen symbol:PRF1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Gene ID:50669
- Antigen synonyms:lymphocyte pore-forming protein|cytolysin|lymphocyte pore forming protein|perforin-1
- Storage buffer:PBS (pH 7,2) containing 30% glycerol, 0,5% BSA, 0,01% thimerosal
- Storage temperature:Aliquot and store at –20 °C. The antibody solution should be gently mixed before use.
- Concentration:0,5 mg/ml
- Purification:Antigen affinity purification
- Size:100 μg
- Pk:100 µG
Type: Primary
Antigen: PRF1
Clonality: Polyclonal
Clone: Unconjugated
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat