Anti-alpha beta - Synuclein (synuclein alpha (non A4 component of amyloid Precursor)) Mouse Monoclonal Antibody [clone: 3B6]
Catalog # GNTXGTX50063
Supplier: Genetex
Specifications
- Antibody type:Primary
- Antigen name:alpha beta - Synuclein (synuclein alpha (non A4 component of amyloid Precursor))
- Clonality:Monoclonal
- Clone:3B6
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Gene ID:6622
- Antigen synonyms:PARK1|PARK4|NACP|PD1
- Storage buffer:Supplied in Phosphate-Buffered Saline (pH 7,4) with 0,1% Sodium Azide.
- Storage temperature:Can be stored at +4C. For long term storage, aliquot and store at -20C. Avoid repeated freezing and thawing cycles.
- Concentration:1 mg/ml
- Immunogen:Recombinant human a-synuclein (119-140aa) purified from E. coli
- Purification:By protein-G affinity chromatography
- Size:100 μl
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen:
Clonality: Monoclonal
Clone: Unconjugated
Conjugation: 3B6
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat