Anti-AFP Mouse Monoclonal Antibody [clone: 131-12210(TB-4B2-10)]
Katalognr. GNTXGTX44462
Anbieter: Genetex
Technische Daten
- Antibody type:Primary
- Antigen name:Alpha 1 Fetoprotein Receptor
- Antigen symbol:AFP
- Clonality:Monoclonal
- Clone:131-12210(TB-4B2-10)
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2b
- Reactivity:Human
- Gene ID:174
- Antigen synonyms:FETA|Alpha-1-Fetoprotein|HPAFP
- Storage buffer:10mM Phosphate, pH 7,2 containing 150mM Sodium chloride
- Storage temperature:Upon receipt, store at –20°C. Avoid multiple freeze/thaw cycles.
- Concentration:5 mg/ml
- Size:1 mg
- Pk:1 mg
Technische Daten
Über diesen Artikel
Type: Primary
Antigen: AFP
Clonality: Monoclonal
Clone: Unconjugated
Conjugation: 131-12210(TB-4B2-10)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human