Anti-Amyloid beta (1-40) (amyloid beta (A4) Precursor Protein) Mouse Monoclonal Antibody [clone: 11A50-B10]
GNTXGTX44423
: Genetex
- Antibody type:Primary
- Antigen name:Amyloid beta (1-40) (amyloid beta (A4) Precursor Protein)
- Clonality:Monoclonal
- Clone:11A50-B10
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Size:100 μL
- Pk:100 µl
Type: Primary
Antigen:
Clonality: Monoclonal
Clone:
Conjugation: 11A50-B10
Epitope:
Host: Mouse
Isotype:
Reactivity: Human