Anti-Amyloid beta (1-42) (amyloid beta (A4) Precursor Protein) Mouse Monoclonal Antibody [clone: BDI780]
Katalognr. GNTXGTX44420
Supplier: Genetex
Specifications
- Antibody type:Primary
- Antigen name:Amyloid beta (1-42) (amyloid beta (A4) Precursor Protein)
- Clonality:Monoclonal
- Clone:BDI780
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Gene ID:351
- Antigen synonyms:CTFgamma|CVAP|ABETA|APPI|AD1|PN-II|ABPP|AAA|PN2
- Storage buffer:PBS with PEG, Sucrose and 0,1% Sodium Azide.
- Storage temperature:Upon receipt, store at –20°C. Avoid multiple freeze/thaw cycles.
- Size:50 μg
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen:
Clonality: Monoclonal
Clone: Unconjugated
Conjugation: BDI780
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human