Anti-APOB Mouse Monoclonal Antibody [clone: 057-10035]
Catalog # GNTXGTX44392
Supplier: Genetex
Specifications
- Antibody type:Primary
- Antigen name:Apolipoprotein B
- Antigen symbol:APOB
- Clonality:Monoclonal
- Clone:057-10035
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG2b
- Reactivity:Human
- Gene ID:338
- Antigen synonyms:apoB-100|apo B-100|FLDB|Apolipoprotein B-100|Apolipoprotein B46|apoB-48|LDLCQ4
- Storage buffer:10mM Phosphate, pH 7,4 containing 150mM Sodium chloride and 0,1% Sodium azide
- Storage temperature:Upon receipt, store at –20°C. Avoid multiple freeze/thaw cycles.
- Size:1 mg
- Pk:1 mg
Specifications
About this item
Type: Primary
Antigen: APOB
Clonality: Monoclonal
Clone: Unconjugated
Conjugation: 057-10035
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human