Anti-BNP and proBNP in immune Complex with H86245M Mouse Monoclonal Antibody () [clone: Ab-BNP2]
Katalognr. GNTXGTX44338
Supplier: Genetex
Specifications
- Antibody type:Primary
- Antigen name:BNP And proBNP in immune Complex with H86245M
- Antigen symbol:BNP and proBNP in immune Complex with H86245M
- Clonality:Monoclonal
- Clone:Ab-BNP2
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Size:200 μg
- Pk:200 µG
Specifications
About this item
Type: Primary
Antigen: BNP and proBNP in immune Complex with H86245M
Clonality: Monoclonal
Clone:
Conjugation: Ab-BNP2
Epitope:
Host: Mouse
Isotype:
Reactivity: Human