Anti-c-myc (v-myc myelocytomatosis viral oncogene homolog (avian)) Mouse Monoclonal Antibody [clone: 289-19510(9E10-F5)]
Catalog # GNTXGTX44310
Supplier: Genetex
Specifications
- Antibody type:Primary
- Antigen name:V-Myc Avian Myelocytomatosis Viral Oncogene Homolog
- Antigen symbol:c-myc (v-myc myelocytomatosis viral oncogene homolog (avian))
- Clonality:Monoclonal
- Clone:289-19510(9E10-F5)
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Gene ID:4609
- Antigen synonyms:MRTL|bHLHe39|c-Myc|MYCC
- Storage buffer:10 mM Phosphate, pH 7,4 containing 150 mM Sodium Chloride and 0,1% Sodium Azide
- Storage temperature:Upon receipt, store at –20°C. Avoid multiple freeze/thaw cycles.
- Size:200 μg
- Pk:200 µG
Specifications
About this item
Type: Primary
Antigen: c-myc (v-myc myelocytomatosis viral oncogene homolog (avian))
Clonality: Monoclonal
Clone: Unconjugated
Conjugation: 289-19510(9E10-F5)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human