Anti-CD41 (Integirn alpha 2b (platelet glycoprotein IIb of IIb/IIIa Complex Antigen CD41)) Mouse Monoclonal Antibody [clone: P2]
Katalognr. GNTXGTX44204
Supplier: Genetex
Specifications
- Antibody type:Primary
- Antigen name:Integrin Alpha 2 Beta
- Antigen symbol:ITGA2B
- Clonality:Monoclonal
- Clone:P2
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Human
- Gene ID:3674
- Antigen synonyms:PPP1R93|GP2B|GTA|CD41B|HPA3|GPIIb|BDPLT2|CD41|BDPLT16|GT
- Storage buffer:Lyophilized from PBS with 1 mg/mL BSA.
- Storage temperature:Upon receipt, store at –20°C. Avoid multiple freeze/thaw cycles.
- Size:100 μg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: CD41 (Integirn alpha 2b (platelet glycoprotein IIb of IIb/IIIa Complex Antigen CD41))
Clonality: Monoclonal
Clone: Unconjugated
Conjugation: P2
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human