Anti-F3 Mouse Monoclonal Antibody (FITC (Fluorescein isothiocyanate)) [clone: CLB/TF-5]
Catalog # GNTXGTX43752
Supplier: Genetex
Specifications
- Antibody type:Primary
- Antigen name:Coagulation Factor 3
- Antigen symbol:F3
- Clonality:Monoclonal
- Clone:CLB/TF-5
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Gene ID:2152
- Antigen synonyms:F3|TFA|Coagulation factor 3|CD142|TF
- Storage buffer:TRIS buffered saline containing 0,09% Sodium Azide (NaN3) 1% Bovine Serum Albumin
- Storage temperature:Store at +4 °C or at –20 °C if preferred. Storage in frost-free freezers is not recommended. Store undiluted. Photosensitive. Should this product contain a precipitate we recommend microcentrifugation before use.
- Purification:Purified IgG prepared by affinity chromatography on Protein A from tissue culture supernatant
- Size:100 μg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: F3
Clonality: Monoclonal
Clone: FITC (Fluorescein isothiocyanate)
Conjugation: CLB/TF-5
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human