Anti-CD16/CD56 (Fc fragment of IgG, low affinity IIIb,R (CD16b)) Mouse Monoclonal Antibody (FITC (Fluorescein isothiocyanate)) [clone: LNK16/MEM-188]
Katalognr. GNTXGTX43737
Supplier: Genetex
Specifications
- Antibody type:Primary
- Antigen name:Neural Cell Adhesion Molecule 1
- Antigen symbol:NCAM1
- Clonality:Monoclonal
- Clone:LNK16/MEM-188
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Gene ID:4684
- Antigen synonyms:NCAM|MSK39|CD56
- Storage buffer:Phosphate buffered saline containing 0,09% Sodium Azide 1% Bovine Serum Albumin
- Storage temperature:Store at +4 °C or at –20 °C if preferred. Store undiluted. Storage in frost free freezers is not recommended. Photosensitive. Should this product contain a precipitate we recommend microcentrifugation before use.
- Purification:Purified IgG prepared by affinity chromatography on Protein G
- Size:25 μg
- Pk:25 µG
Specifications
About this item
Type: Primary
Antigen: CD16/CD56 (Fc fragment of IgG, low affinity IIIb,R (CD16b))
Clonality: Monoclonal
Clone: FITC (Fluorescein isothiocyanate)
Conjugation: LNK16/MEM-188
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human