Anti-FCE2 Mouse Monoclonal Antibody (FITC (Fluorescein isothiocyanate)) [clone: Bu38]
GNTXGTX43667
: Genetex
- Antibody type:Primary
- Antigen name:Fc fragment of IgE receptor 2
- Antigen symbol:FCE2
- Clonality:Monoclonal
- Clone:Bu38
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Gene ID:2208
- Antigen synonyms:CLEC4J|BLAST-2|FCE2|CD23A|CD23|IGEBF
- Storage buffer:Phosphate buffered saline containing 0,09% Sodium Azide (NaN3) 1% Bovine Serum Albumin
- Storage temperature:Store at +4 °C or at –20 °C if preferred. Storage in frost-free freezers is not recommended. Store undiluted. Photosensitive. Should this product contain a precipitate we recommend microcentrifugation before use.
- Purification:Purified IgG prepared by affinity chromatography on Protein G from tissue culture supernatant
- Size:100 μg
- Pk:100 µG
Type: Primary
Antigen: FCE2
Clonality: Monoclonal
Clone: FITC (Fluorescein isothiocyanate)
Conjugation: Bu38
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human