Anti-CD36 Mouse Monoclonal Antibody (FITC (Fluorescein isothiocyanate)) [clone: SM0]
Catalog # GNTXGTX43613
Supplier: Genetex
Specifications
- Antibody type:Primary
- Antigen name:CD36 molecule
- Antigen symbol:CD36
- Clonality:Monoclonal
- Clone:SM0
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgM
- Reactivity:Human
- Gene ID:948
- Antigen synonyms:GP4|GP3B|FAT|GPIV|SCARB3|CHDS7|BDPLT10|PASIV
- Storage buffer:Phosphate buffered saline containing 0,09% Sodium Azide 1% Bovine Serum Albumin
- Storage temperature:Store at +4 °C. DO NOT FREEZE. This product should be stored undiluted. This product is photosensitive and should be protected from light. Should this product contain a precipitate we recommend microcentrifugation before use.
- Purification:Purified IgM prepared by ammonium sulphate precipitation from tissue culture supernatant
- Size:25 μg
- Pk:25 µG
Specifications
About this item
Type: Primary
Antigen: CD36
Clonality: Monoclonal
Clone: FITC (Fluorescein isothiocyanate)
Conjugation: SM0
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human