Anti-PTPRC Mouse Monoclonal Antibody (FITC (Fluorescein isothiocyanate)) [clone: Bu78]
Catalog # GNTXGTX43576
Supplier: Genetex
Specifications
- Antibody type:Primary
- Antigen name:Protein tyrosine phosphatase receptor type C
- Antigen symbol:PTPRC
- Clonality:Monoclonal
- Clone:Bu78
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Gene ID:5788
- Antigen synonyms:CD45R|B220|CD45|GP180|T200|L-CA|LY5|receptor type C|LCA|protein tyrosine phosphatase
- Storage buffer:Phosphate buffered saline containing 0,09% Sodium Azide (NaN3) 1% Bovine Serum Albumin
- Storage temperature:Store at +4 °C or at –20 °C if preferred. Storage in frost-free freezers is not recommended. Store undiluted. Photosensitive. Should this product contain a precipitate we recommend microcentrifugation before use.
- Purification:Purified IgG prepared by affinity chromatography on Protein G from tissue culture supernatant
- Size:100 μg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: PTPRC
Clonality: Monoclonal
Clone: FITC (Fluorescein isothiocyanate)
Conjugation: Bu78
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human