Anti-CD9 Mouse Monoclonal Antibody (FITC (Fluorescein isothiocyanate)) [clone: Bu16]
GNTXGTX43478
: Genetex
- Antibody type:Primary
- Antigen name:CD9 molecule
- Antigen symbol:CD9
- Clonality:Monoclonal
- Clone:Bu16
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Human
- Gene ID:928
- Antigen synonyms:MIC3|TSPAN29|TSPAN-29|DRAP-27|MRP-1|BTCC-1
- Storage buffer:Phosphate buffered saline containing 0,09% Sodium Azide (NaN3) 1% Bovine Serum Albumin
- Storage temperature:Store at +4 °C or at –20 °C if preferred. Storage in frost-free freezers is not recommended. Store undiluted. Photosensitive. Should this product contain a precipitate we recommend microcentrifugation before use.
- Purification:Purified IgG prepared by affinity chromatography on Protein G from tissue culture supernatant
- Size:100 μg
- Pk:100 µG
Type: Primary
Antigen: CD9
Clonality: Monoclonal
Clone: FITC (Fluorescein isothiocyanate)
Conjugation: Bu16
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human