Anti-CTL1 Mouse Monoclonal Antibody (FITC (Fluorescein isothiocyanate)) [clone: VIM-15b]
Catalog # GNTXGTX43476
Supplier: Genetex
Specifications
- Antibody type:Primary
- Antigen name:Choline transporter-like protein 1
- Antigen symbol:CTL1
- Clonality:Monoclonal
- Clone:VIM-15b
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG2b
- Reactivity:Human
- Gene ID:23446
- Antigen synonyms:CTL1|CD92|CDW92|solute carrier family 44 member 1|CHTL1
- Storage buffer:Phosphate buffered saline containing 0,09% Sodium Azide (NaN3) 1% Bovine Serum Albumin
- Storage temperature:Store at +4 °C or at –20 °C if preferred. Storage in frost-free freezers is not recommended. Store undiluted. Photosensitive. Should this product contain a precipitate we recommend microcentrifugation before use.
- Purification:Purified IgG prepared by affinity chromatography on Protein G from tissue culture supernatant
- Size:100 μg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: CTL1
Clonality: Monoclonal
Clone: FITC (Fluorescein isothiocyanate)
Conjugation: VIM-15b
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human