Anti-ZAP 70 Mouse Monoclonal Antibody (FITC (Fluorescein isothiocyanate)) [clone: SBZAP]
Catalog # GNTXGTX43323
Supplier: Genetex
Specifications
- Antibody type:Primary
- Antigen name:Zeta-chain (TCR) associated Protein kinase 70kDa
- Antigen symbol:ZAP 70
- Clonality:Monoclonal
- Clone:SBZAP
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Gene ID:7535
- Antigen synonyms:ZAP-70|SRK|TZK|STD|STCD
- Storage buffer:Phosphate buffered saline containing 0,1% Sodium azide
- Storage temperature:Store at +4 °C or at –20 °C if preferred. Storage in frost-free freezers is not recommended. Store undiluted. Photosensitive. Should this product contain a precipitate we recommend microcentrifugation before use.
- Size:100 μg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: ZAP 70
Clonality: Monoclonal
Clone: FITC (Fluorescein isothiocyanate)
Conjugation: SBZAP
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human