Anti-DEFB1 Mouse Monoclonal Antibody [clone: M11-14b-D10]
GNTXGTX43303
: Genetex
- Antibody type:Primary
- Antigen name:Defensin beta-1 (defensin beta 1)
- Antigen symbol:DEFB1
- Clonality:Monoclonal
- Clone:M11-14b-D10
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Gene ID:1672
- Antigen synonyms:HBD1|DEFB101|BD1|DEFB-1
- Storage buffer:Lyophilized from PBS, pH 7,4. and
- Storage temperature:Upon receipt, store at –20°C. Avoid multiple freeze/thaw cycles.
- Size:10 μg
- Pk:10 µG
Type: Primary
Antigen: DEFB1
Clonality: Monoclonal
Clone: Unconjugated
Conjugation: M11-14b-D10
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human