Anti-AMPK alpha 1/2 Mouse Monoclonal Antibody [clone: 84.1]
Catalog # GNTXGTX42788
Supplier: Genetex
Specifications
- Antibody type:Primary
- Antigen name:5' AMP-activated protein kinase alpha 1 And 2
- Antigen symbol:AMPK alpha 1/2
- Clonality:Monoclonal
- Clone:84.1
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG2a
- Reactivity:Drosophila
- Western blot:Yes
- Environmentally Preferable:
- Gene ID:5562
- Antigen synonyms:AMPK|AMPKa1
- Storage buffer:Phosphate buffered saline containing 0,09% Sodium Azide (NaN3)
- Storage temperature:Store at +4 °C or at –20 °C if preferred. Storage in frost-free freezers is not recommended. This product should be stored undiluted. Should this product contain a precipitate we recommend microcentrifugation before use.
- Immunogen:29 amino acid peptide within the N-terminal catalytic domain of Drosophila AMPK alpha.
- Purification:Purified IgG prepared by affinity chromatography on Protein G
- Size:100 μg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: AMPK alpha 1/2
Clonality: Monoclonal
Clone: Unconjugated
Conjugation: 84.1
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Drosophila