Anti-CD75s (ST6 beta-galactosamide alpha-2 6-sialyltranferase 1) Mouse Monoclonal Antibody [clone: LN-1]
GNTXGTX41842
: Genetex
- Antibody type:Primary
- Antigen name:ST6 beta-galactosamide alpha-2,6-sialyltranferase 1
- Antigen symbol:ST6GAL1
- Clonality:Monoclonal
- Clone:LN-1
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgM
- Reactivity:Pig,Human,Mink
- Gene ID:6480
- Antigen synonyms:ST6GalI|ST6N|SIAT1
- Storage buffer:Phosphate buffered saline containing 0,05% Sodium Azide
- Storage temperature:Store at +4 °C or at –20 °C if preferred. This product should be stored undiluted. Storage in frost-free freezers is not recommended. Should this product contain a precipitate we recommend microcentrifugation before use.
- Size:100 μg
- Pk:100 µG
Type: Primary
Antigen: CD75s (ST6 beta-galactosamide alpha-2 6-sialyltranferase 1)
Clonality: Monoclonal
Clone: Unconjugated
Conjugation: LN-1
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human, Pig, Mink