Anti-INHA Mouse Monoclonal Antibody [clone: R1]
GNTXGTX40215
: Genetex
- Antibody type:Primary
- Antigen name:Inhibin Alpha
- Antigen symbol:INHA
- Clonality:Monoclonal
- Clone:R1
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2a
- Reactivity:Sheep,Human,Bovine
- Gene ID:3623
- Antigen synonyms:RIR1|AW555078|RR1|R1
- Immunogen:Synthetic peptide comprised of amino acids 1-32 of human inhibin alpha.
- Purification:Tissue Culture Supernatant
- Size:200 μl
- Pk:200 µl
Type: Primary
Antigen: INHA
Clonality: Monoclonal
Clone: Unconjugated
Conjugation: R1
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Sheep, Bovine