Anti-RLN2 Mouse Monoclonal Antibody [clone: 8H11kurd]
Catalog # GNTXGTX39294
Supplier: Genetex
Specifications
- Antibody type:Primary
- Antigen name:Relaxin 2
- Antigen symbol:RLN2
- Clonality:Monoclonal
- Clone:8H11kurd
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Gene ID:6019
- Antigen synonyms:bA12D24.1.1|RLXH2|bA12D24.1.2|H2|H2-RLX
- Storage buffer:Lyophilized from PBS, pH 7,4 and 0,1% Sodium azide (prior to lyophilization)
- Storage temperature:Upon receipt, store at –20°C. Avoid multiple freeze/thaw cycles.
- Size:10 μg
- Pk:10 µG
Specifications
About this item
Type: Primary
Antigen: RLN2
Clonality: Monoclonal
Clone: Unconjugated
Conjugation: 8H11kurd
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human