Anti-MFG Mouse Monoclonal Antibody () [clone: MFG-06]
Catalog # GNTXGTX22537
Supplier: Genetex
Specifications
- Antibody type:Primary
- Antigen symbol:MFG
- Clonality:Monoclonal
- Clone:MFG-06
- Host:Mouse
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Size:100 μg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: MFG
Clonality: Monoclonal
Clone:
Conjugation: MFG-06
Epitope:
Host: Mouse
Isotype:
Reactivity: Human