Anti-CG beta (chorionic gonadotropin, beta Polypeptide) Mouse Monoclonal Antibody [clone: HCG-61]
Catalog # GNTXGTX20764
Supplier: Genetex
Specifications
- Antibody type:Primary
- Antigen name:Chorionic Gonadotropin, Beta Polypeptide
- Antigen symbol:CG beta (chorionic gonadotropin, beta Polypeptide)
- Clonality:Monoclonal
- Clone:HCG-61
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG2b
- Reactivity:Human
- Gene ID:1082
- Antigen synonyms:CGB3|CGB8|CGB5|hCGB|CGB7
- Storage buffer:Phosphate buffered saline pH 7,4 containing 15mM sodium azide
- Storage temperature:Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:Full length native protein (purified) (Human).
- Purification:>95% by SDS-PAGE
- Size:100 μg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: CG beta (chorionic gonadotropin, beta Polypeptide)
Clonality: Monoclonal
Clone: Unconjugated
Conjugation: HCG-61
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human