Order Entry
Switzerland
ContactUsLinkComponent
Anti-CD1C Mouse Monoclonal Antibody [clone: M241]
Anti-CD1C Mouse Monoclonal Antibody [clone: M241]
  GNTXGTX18216
 :  Genetex
Anti-CD1C Mouse Monoclonal Antibody [clone: M241]
  GNTXGTX18216
 :  Genetex

 

  • Antibody type:
    Primary
  • Antigen name:
    CD1c molecule
  • Antigen symbol:
    CD1C
  • Clonality:
    Monoclonal
  • Clone:
    M241
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Gene ID:
    911
  • Antigen synonyms:
    BDCA1|CD1A|T-cell surface glycoprotein CD1c|CD1|RP11-101J8.3|differentiation antigen CD1-alpha-3|R7|c polypeptide|cortical thymocyte antigen CD1C|CD1C antigen
  • Storage buffer:
    Preservative: None. Constituents: PBS
  • Storage temperature:
    Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
  • Concentration:
    1 mg/ml
  • Immunogen:
    MOLT 4 cell line.
  • Purification:
    IgG fraction
  • Size:
    100 μg
  • Pk:
    100 µG

 

 

Type: Primary
Antigen: CD1C
Clonality: Monoclonal
Clone: Unconjugated
Conjugation: M241
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human