Anti-LIMP2 Mouse Monoclonal Antibody () [clone: JDR.3B8]
GNTXGTX11313
: Genetex
- Antibody type:Primary
- Antigen name:Lysosome membrane protein 2
- Antigen symbol:LIMP2
- Clonality:Monoclonal
- Clone:JDR.3B8
- Host:Mouse
- ImmunoFluorescence:Yes
- Reactivity:Human,Mouse
- Western blot:Yes
- Antigen synonyms:CD36L2|AMRF|scavenger receptor class B|member 2|LIMPII|85 kDa lysosomal sialoglycoprotein scavenger receptor class B|CD36 antigen (collagen type I receptor|thrombospondin receptor)-like 2 (lysosomal integral membrane protein II)|SCARB2|lysosomal integral membrane protein II|HLGP85
- Size:100 μL
- Pk:100 µl
Type: Primary
Antigen: LIMP2
Clonality: Monoclonal
Clone:
Conjugation: JDR.3B8
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse