Anti-ATXN2 Mouse Monoclonal Antibody (Biotin) [clone: 202/7B1]
Catalog # GNTXGTX11206
Supplier: Genetex
Specifications
- Antibody type:Primary
- Antigen name:Ataxin 2
- Antigen symbol:ATXN2
- Clonality:Monoclonal
- Clone:202/7B1
- Conjugation:Biotin
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Human
- Gene ID:1241
- Antigen synonyms:LTBR1|LTB4R1|GPR16|CMKRL1|P2Y7|P2RY7|BLT1|BLTR
- Storage buffer:Phosphate buffered saline containing 0,09% Sodium Azide 1% Bovine Serum Albumin
- Purification:Purified IgG prepared by affinity chromatography on Protein G from tissue culture supernatant
- Size:50 μg
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: ATXN2
Clonality: Monoclonal
Clone: Biotin
Conjugation: 202/7B1
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human