Order Entry
Switzerland
ContactUsLinkComponent
Anti-XRCC1 Rabbit Polyclonal Antibody
Anti-XRCC1 Rabbit Polyclonal Antibody
Catalog # GNTXGTX111712
Supplier:  Genetex
Anti-XRCC1 Rabbit Polyclonal Antibody
Catalog # GNTXGTX111712
Supplier:  Genetex

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    X-ray Repair Complementing Defective Repair in Chinese cells 1
  • Antigen symbol:
    XRCC1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat
  • Western blot:
    Yes
  • Gene ID:
    7515
  • Antigen synonyms:
    complementing defective|X-ray-repair|X-ray repair cross-complementing protein 1|DNA repair protein XRCC1|repair in Chinese hamster
  • Storage buffer:
    0,1M Tris, 0,1M Glycine, 10% Glycerol (pH7). 0,01% Thimerosal was added as a preservative.
  • Molecular weight:
    70 kDa
  • Storage temperature:
    Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid multiple freeze-thaw cycles.
  • Concentration:
    0,73 mg/ml
  • Immunogen:
    Recombinant protein encompassing a sequence within the center region of human XRCC1. The exact sequence is proprietary.
  • Tested applications:
    ICC/IF, IHC-P, WB
  • Purification:
    Purified by antigen-affinity chromatography.
  • Size:
    100 μl
  • Pk:
    25 µl

Specifications

About this item

Type: Primary
Antigen: XRCC1
Clonality: Polyclonal
Clone: Unconjugated
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat