Order Entry
Switzerland
ContactUsLinkComponent
Anti-CCND1 Mouse Monoclonal Antibody (CF640R) [clone: DCS-6]
Anti-CCND1 Mouse Monoclonal Antibody (CF640R) [clone: DCS-6]
Supplier:  Biotium
Certificates
Anti-CCND1 Mouse Monoclonal Antibody (CF640R) [clone: DCS-6]
Supplier:  Biotium

Specifications

  • Catalog No:
  • BNC400007-500
  • BNC400007-100
  • Antigen symbol:
  • CCND1
  • CCND1
  • Antigen name:
  • Cyclin D1
  • Cyclin D1
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 640R
  • CF® 640R
  • Clone:
  • Clone DCS-6
  • Clone DCS-6
  • Reactivity:
  • Monkey, Human, Rat, Mouse
  • Monkey, Human, Rat, Mouse
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 523852 , 667996
  • 523852 , 667996
  • Isotype:
  • IgG2a kappa
  • IgG2a kappa
  • Western blot:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

Specifications

Frequently Bought Together