Order Entry
Switzerland
ContactUsLinkComponent
VWR®, Uranium standard solutions for ICP-MS, ARISTAR®
VWR®, Uranium standard solutions for ICP-MS, ARISTAR®
Catalog # 85641.180
Supplier:  VWR Chemicals
CAS Number:  
undefined
VWR®, Uranium standard solutions for ICP-MS, ARISTAR®
Catalog # 85641.180
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Concentration:
    10 ppm
  • Matrix:
    2% HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Seq:
    141
  • Element:
    Uranium
  • Symbol:
    U

Specifications

About this item

  • Produced from high purity acids, water ASTM I 18 Megaohms
  • Solution assayed by titration
  • Final concentration verified against reference material from NIST
  • Total maximum uncertainty: 0,5% (typical)
  • Delivered with complete certificate of analysis
Certifications: Manufactured and tested in ISO 17034 / ISO 17025 accredited facilities