VWR® ARISTAR®, Phosphorus Standard Solutions for ICP-MS
Supplier: VWR Chemicals
Certificates
About this item
- Produced from high purity acids, water ASTM I 18 Megaohms
- Solution assayed by titration
- Final concentration verified against reference material from NIST
- Total maximum uncertainty: 0.5% (typical)
- Delivered with complete certificate of analysis
Certifications: Manufactured and tested in ISO 17034 / ISO 17025 accredited facilities
Limited Time Sale
Select items below are currently on sale
Related Information
Certificates
Search by Lot Number125 ml Plastic bottle for liquids DIN 24 (ZPACK15150125)
Packaging Information
VWR® for Calibrations
Product Literature
Frequently Bought Together