Anti-RICK Rabbit Polyclonal Antibody
ENZOADIAAP460E
- Antibody type:Primary
- Antigen name:Receptor-interacting serine-threonine kinase 2
- Antigen symbol:RIPK2
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Western blot:Yes
- Seq:02
- Epitope:near the N-Terminus
- Form:Liquid
- Antigen synonyms:CARD3|RICK|CARDIAK|receptor interacting serine/threonine kinase 2|GIG30|CCK|RIP2
- Storage buffer:PBS containing 0,02% sodium azide
- Storage temperature:4 °C
- Shipping temperature:blue ice not frozen
- Immunogen:Synthetic peptide corresponding to the sequence near the N-terminus of human RICK
- Purification:Purified by peptide affinity chromatography
- Size:100 μg
- Pk:100 µG
The RICK (Receptor-interacting serine/threonine protein kinase 2) protein, also known as RIP-like interacting CLARP kinase, and CARD-containing ICE associated kinase, contains an N-terminal kinase domain and a C-terminal CARD domain.
- Dilution range: Immunocytochemistry (10 µg/ml); Western blot (0,5 to 1,0 µg/ml, ECL)
Overexpression of RICK results in the activation of NF-κB pathway and also induces apoptosis. RICK represents a kinase that regulates TNF and Fas induced-apoptosis and that is involved in the generation of proinflammatory cytokine IL-1beta.
Type: Primary
Antigen: Receptor-interacting serine/threonine-protein kinase 2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope: N-Terminal
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat