Autoclavable goggles, BCAH series, BioClean Clearview™
NITRBCAH
: Nitritex
- Type:Cleanroom, autoclavable, anti-scratch
- Colour:White
- Description:Autoclavable goggles
- Lenses:Clear PC
- Pk:12
BioClean Clearview™ autoclavable cleanroom goggles have a super-soft thermoplastic rubber frame to provide wearer comfort.
- Anti-fog lens
- Super-soft frame
- Indirect ventilation system
- Non-linting, latex-free headband
- Tested to withstand 40 autoclave cycles of 30 minutes duration at 121 °C/250 °F (under laboratory conditions)
- Anti-fog performance remaining for up to 25 cycles with no signs of degradation (under laboratory conditions)
Constructed from super-soft, lightweight silicone rubber to provide user comfort and enable prolonged use, BioClean Clearview™ Autoclavable Cleanroom Goggles feature an indirect ventilation system to reduce the risk of contamination entering the controlled environment, and have an anti-fog polycarbonate lens.
: Manufactured in a facility holding ISO 9001:2015. Processed in an NEBB certified ISO Class 4 cleanroom environment.
: Goggle (BCAH): One piece per sealed PE bag (to be removed prior to autoclaving); 12 bags per inner box; five boxes per lined carton (60 pieces). Replacement lens (BCAH-SL): One piece per sealed PE bag; 10 bags per inner box; 10 boxes per lined carton (100 pieces). Replacement strap (BCAH-SS): 25 pieces per sealed PE bag; two bags per inner box; 10 boxes per lined carton (500 pieces).